Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097077.1 | XP_020952736.1 | 214 | 76 | ||
| GenBank mRNA | AY610321.1 | - | 2 - 3 | 114 | 76 | |
More Information
Set Membership: |
|

DYNLRB2
Genomic Map