Assay Details
| Gene Symbol: POP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013995472.2 | XP_013850926.1 | 667 | 92 | ||
| GenBank mRNA | AY610322.1 | - | 1 - 2 | 165 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013995472.2 transcripts.
Set Membership: |
|

POP7
Genomic Map