Assay Details
| Gene Symbol: LOC100738180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D49580.1 | - | 1 - 2 | 124 | 106 | |
| D49581.1 | - | 1 - 2 | 118 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D49580.1, D49581.1 transcripts.
Set Membership: |
|

LOC100738180
Genomic Map