Assay Details
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080957.1 | XP_020936616.1 | 1608 | 94 | ||
| GenBank mRNA | DQ139314.1 | - | 1 - 2 | 139 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: STS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080957.1 | XP_020936616.1 | 1608 | 94 | ||
| GenBank mRNA | DQ139314.1 | - | 1 - 2 | 139 | 94 | |
Set Membership: |
|