Assay Details
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001367266.1 | NP_001354195.1 | 2 - 3 | 267 | 77 | |
GenBank mRNA | EU871629.1 | - | 1 - 2 | 73 | 77 | |
FJ588029.1 | - | 2 - 3 | 267 | 77 | ||
HG518672.1 | - | 1 - 2 | 112 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
