Assay Details
| Gene Symbol: PGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001925764.5 | XP_001925799.1 | 687 | 76 | ||
| XM_003356751.4 | XP_003356799.1 | 707 | 76 | |||
| GenBank mRNA | FJ177137.1 | - | 3 - 4 | 360 | 76 | |
More Information
Set Membership: |
|

PGF
Genomic Map