Assay Details
| Gene Symbol: KCNMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091986.1 | XP_020947645.1 | 416 | 123 | ||
| GenBank mRNA | FJ645572.1 | - | 1 - 2 | 285 | 123 | |
More Information
Set Membership: |
|

KCNMB4
Genomic Map