Assay Details
| Gene Symbol: CST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077681.1 | XP_020933340.1 | 308 | 82 | ||
| GenBank mRNA | FM995238.1 | - | 1 - 2 | 179 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077681.1 | XP_020933340.1 | 308 | 82 | ||
| GenBank mRNA | FM995238.1 | - | 1 - 2 | 179 | 82 | |
Set Membership: |
|