Assay Details
| Gene Symbol: WFDC10AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134467.4 | XP_003134515.1 | 184 | 96 | ||
| GenBank mRNA | FM995241.1 | - | 1 - 2 | 98 | 96 | |
More Information
Set Membership: |
|

WFDC10AL
Genomic Map