Assay Details
| Gene Symbol: WFDC10AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134467.4 | XP_003134515.1 | 337 | 72 | ||
| GenBank mRNA | FM995241.1 | - | 2 - 3 | 251 | 72 | |
More Information
Set Membership: |
|

WFDC10AL
Genomic Map