Assay Details
| Gene Symbol: CNNM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087271.1 | XP_020942930.1 | 1650 | 70 | ||
| XM_021087272.1 | XP_020942931.1 | 1650 | 70 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CNNM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087271.1 | XP_020942930.1 | 1650 | 70 | ||
| XM_021087272.1 | XP_020942931.1 | 1650 | 70 | |||
Set Membership: |
|