Assay Details
| Gene Symbol: FAM168B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075749.1 | XP_020931408.1 | 890 | 90 | ||
| XM_021075750.1 | XP_020931409.1 | 722 | 90 | |||
| XM_021075751.1 | XP_020931410.1 | 752 | 90 | |||
More Information
Set Membership: |
|

FAM168B
Genomic Map