Assay Details
| Gene Symbol: KLHL24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132561.5 | XP_003132609.2 | 1500 | 65 | ||
| XM_005652345.3 | XP_005652402.1 | 1584 | 65 | |||
| XM_005652348.3 | XP_005652405.1 | 1447 | 65 | |||
| XM_005652349.3 | XP_005652406.1 | 1347 | 65 | |||
| XM_005652350.3 | XP_005652407.1 | 1450 | 65 | |||
| XM_013980505.2 | XP_013835959.1 | 1524 | 65 | |||
| XM_013981678.2 | XP_013837132.1 | 1323 | 65 | |||
| XM_021069901.1 | XP_020925560.1 | 1622 | 65 | |||
| XM_021069902.1 | XP_020925561.1 | 2086 | 65 | |||
| XM_021069903.1 | XP_020925562.1 | 1517 | 65 | |||
| XM_021069905.1 | XP_020925564.1 | 1560 | 65 | |||
| XM_021069906.1 | XP_020925565.1 | 1580 | 65 | |||
| XM_021069907.1 | XP_020925566.1 | 1475 | 65 | |||
| XM_021069908.1 | XP_020925567.1 | 715 | 65 | |||
More Information
Set Membership: |
|

KLHL24
Genomic Map