Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080829.1 | XP_020936488.1 | 2480 | 61 | ||
| XM_021080830.1 | XP_020936489.1 | 2511 | 61 | |||
| XM_021080831.1 | XP_020936490.1 | 2294 | 61 | |||
More Information
Set Membership: |
|

TAB3
Genomic Map