Assay Details
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134871.6 | XP_003134919.2 | 6176 | 73 | ||
| XM_021079068.1 | XP_020934727.1 | 5895 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134871.6 | XP_003134919.2 | 6176 | 73 | ||
| XM_021079068.1 | XP_020934727.1 | 5895 | 73 | |||
Set Membership: |
|