Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094669.1 | XP_020950328.1 | 269 | 67 | ||
XM_021094670.1 | XP_020950329.1 | 550 | 67 | |||
XM_021094671.1 | XP_020950330.1 | 547 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
