Assay Details
| Gene Symbol: LRRC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981331.2 | XP_013836785.1 | 2895 | 102 | ||
| XM_021072980.1 | XP_020928639.1 | 2559 | 102 | |||
More Information
Set Membership: |
|

LRRC43
Genomic Map