Assay Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673830.3 | XP_005673887.1 | 3765 | 101 | ||
XM_005673831.3 | XP_005673888.1 | 571 | 101 | |||
XM_013986332.2 | XP_013841786.1 | 641 | 101 | |||
XM_013986334.2 | XP_013841788.1 | 667 | 101 | |||
XM_013986335.2 | XP_013841789.1 | 641 | 101 | |||
XM_013986336.2 | XP_013841790.1 | 640 | 101 | |||
XM_013986337.2 | XP_013841791.1 | 640 | 101 | |||
XM_013986338.2 | XP_013841792.1 | 640 | 101 | |||
XM_013986339.2 | XP_013841793.1 | 640 | 101 | |||
XM_021080436.1 | XP_020936095.1 | 687 | 101 | |||
XM_021080437.1 | XP_020936096.1 | 3764 | 101 | |||
XM_021080438.1 | XP_020936097.1 | 3763 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
