Assay Details
| Gene Symbol: CTDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660586.3 | XP_005660643.2 | 2352 | 52 | ||
| XM_013993984.2 | XP_013849438.2 | 2101 | 52 | |||
| XM_013993985.2 | XP_013849439.2 | 2325 | 52 | |||
More Information
Set Membership: |
|

CTDP1
Genomic Map