Assay Details
| Gene Symbol: SMPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021062347.1 | XP_020918006.1 | 1487 | 71 | ||
| GenBank mRNA | AK232225.1 | - | 3 - 4 | 533 | 71 | |
More Information
Set Membership: |
|

SMPD1
Genomic Map