Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480672.4 | XP_003480720.1 | 890 | 62 | ||
| XM_021082805.1 | XP_020938464.1 | 1106 | 62 | |||
| GenBank mRNA | AK232258.1 | - | 9 - 10 | 850 | 62 | |
| AK233584.1 | - | 9 - 10 | 826 | 62 | ||
| AK392644.1 | - | 9 - 10 | 870 | 62 | ||
More Information
Set Membership: |
|

BRMS1
Genomic Map