Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655871.3 | XP_005655928.1 | 1299 | 81 | ||
| XM_005655872.3 | XP_005655929.1 | 1218 | 81 | |||
| XM_013988612.2 | XP_013844066.1 | 727 | 81 | |||
| XM_021094422.1 | XP_020950081.1 | 662 | 81 | |||
| GenBank mRNA | AK232262.1 | - | 4 - 5 | 530 | 81 | |
| AK351140.1 | - | 4 - 5 | 510 | 81 | ||
| AK393164.1 | - | 5 - 6 | 602 | 81 | ||
| AK395635.1 | - | 4 - 5 | 533 | 81 | ||
| AK396379.1 | - | 5 - 6 | 602 | 81 | ||
| AK396765.1 | - | 4 - 5 | 531 | 81 | ||
| AK396997.1 | - | 7 - 8 | 818 | 81 | ||
| AK398186.1 | - | 4 - 5 | 530 | 81 | ||
| AK400209.1 | - | 7 - 8 | 787 | 81 | ||
| AK400862.1 | - | 4 - 5 | 515 | 81 | ||
More Information
Set Membership: |
|

DMAC2
Genomic Map