Assay Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483499.4 | XP_003483547.1 | 439 | 66 | ||
| GenBank mRNA | AK232293.1 | - | 2 - 3 | 437 | 66 | |
| AY610502.1 | - | 1 - 2 | 274 | 66 | ||
More Information
Set Membership: |
|

MRPS16
Genomic Map