Assay Details
| Gene Symbol: LOC100620470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355459.4 | XP_003355507.1 | 278 | 68 | ||
| GenBank mRNA | AK232303.1 | - | 1 - 2 | 97 | 68 | |
| AK233183.1 | - | 1 - 2 | 104 | 68 | ||
| AK394106.1 | - | 1 - 2 | 133 | 68 | ||
| AK394142.1 | - | 1 - 2 | 133 | 68 | ||
| AK398963.1 | - | 1 - 2 | 210 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003355459.4 transcripts.
Set Membership: |
|

LOC100620470
Genomic Map