Assay Details
| Gene Symbol: LOC100620470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355459.4 | XP_003355507.1 | 604 | 65 | ||
| XM_005663895.3 | XP_005663952.1 | 460 | 65 | |||
| GenBank mRNA | AK232303.1 | - | 2 - 3 | 423 | 65 | |
| AK233183.1 | - | 2 - 3 | 430 | 65 | ||
| AK233628.1 | - | 3 - 4 | 687 | 65 | ||
| AK394106.1 | - | 2 - 3 | 459 | 65 | ||
| AK394142.1 | - | 2 - 3 | 459 | 65 | ||
| AK398963.1 | - | 2 - 3 | 536 | 65 | ||
More Information
Set Membership: |
|

LOC100620470
Genomic Map