Assay Details
| Gene Symbol: LOC100620470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355459.4 | XP_003355507.1 | 1031 | 102 | ||
| XM_005663895.3 | XP_005663952.1 | 887 | 102 | |||
| GenBank mRNA | AK232303.1 | - | 4 - 5 | 850 | 102 | |
| AK233183.1 | - | 4 - 5 | 857 | 102 | ||
| AK233628.1 | - | 5 - 6 | 1114 | 102 | ||
| AK394106.1 | - | 4 - 5 | 886 | 102 | ||
| AK394142.1 | - | 4 - 5 | 886 | 102 | ||
| AK398963.1 | - | 4 - 5 | 102 | |||
More Information
Set Membership: |
|

LOC100620470
Genomic Map