Assay Details
| Gene Symbol: ALDH1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021064911.1 | XP_020920570.1 | 831 | 66 | ||
| GenBank mRNA | AK232310.1 | - | 7 - 8 | 797 | 66 | |
| AK393964.1 | - | 7 - 8 | 797 | 66 | ||
More Information
Set Membership: |
|

ALDH1A1
Genomic Map