Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134861.6 | XP_003134909.1 | 312 | 96 | ||
| GenBank mRNA | AK232423.1 | - | 4 - 5 | 301 | 96 | |
| AK350376.1 | - | 1 - 2 | 54 | 96 | ||
More Information
Set Membership: |
|

UBE2D4
Genomic Map