Assay Details
| Gene Symbol: FUOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927817.6 | XP_001927852.1 | 781 | 76 | ||
| XM_003133245.5 | XP_003133293.1 | 298 | 76 | |||
| XM_021073957.1 | XP_020929616.1 | 298 | 76 | |||
| GenBank mRNA | AK232508.1 | - | 3 - 4 | 246 | 76 | |
| AK346357.1 | - | 2 - 3 | 219 | 76 | ||
| AK391164.1 | - | 3 - 4 | 246 | 76 | ||
| AK398174.1 | - | 3 - 4 | 246 | 76 | ||
More Information
Set Membership: |
|

FUOM
Genomic Map