Assay Details
| Gene Symbol: B4GALT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927335.6 | XP_001927370.3 | 1235 | 95 | ||
| XM_001927348.4 | XP_001927383.1 | 1314 | 95 | |||
| XM_013996833.2 | XP_013852287.1 | 1121 | 95 | |||
| XM_013996834.2 | XP_013852288.1 | 1297 | 95 | |||
| XM_021089531.1 | XP_020945190.1 | 1263 | 95 | |||
| XM_021089532.1 | XP_020945191.1 | 1669 | 95 | |||
| GenBank mRNA | AK232520.1 | - | 2 - 3 | 330 | 95 | |
| AK235562.1 | - | 6 - 7 | 1132 | 95 | ||
| AK347611.1 | - | 1 - 2 | 134 | 95 | ||
| AK391115.1 | - | 6 - 7 | 1050 | 95 | ||
| AK396852.1 | - | 6 - 7 | 1044 | 95 | ||
| AK399159.1 | - | 6 - 7 | 1039 | 95 | ||
More Information
Set Membership: |
|

B4GALT3
Genomic Map