Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124561.6 | XP_003124609.3 | 1426 | 166 | ||
| GenBank mRNA | AK232522.1 | - | 10 - 11 | 1374 | 166 | |
| AK393717.1 | - | 10 - 11 | 1409 | 166 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map