Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124561.6 | XP_003124609.3 | 1043 | 114 | ||
| XM_021086520.1 | XP_020942179.1 | 1033 | 114 | |||
| XM_021086521.1 | XP_020942180.1 | 1042 | 114 | |||
| GenBank mRNA | AK232522.1 | - | 6 - 7 | 991 | 114 | |
| AK393717.1 | - | 6 - 7 | 1026 | 114 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map