Assay Details
| Gene Symbol: PKNOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021071043.1 | XP_020926702.1 | 8698 | 56 | ||
| GenBank mRNA | AK232573.1 | - | 2 - 3 | 303 | 56 | |
| AK235324.1 | - | 5 - 6 | 733 | 56 | ||
More Information
Set Membership: |
|

PKNOX1
Genomic Map