Assay Details
| Gene Symbol: SIAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358611.4 | XP_003358659.1 | 868 | 112 | ||
| GenBank mRNA | AK232587.1 | - | 1 - 2 | 718 | 112 | |
| AK350525.1 | - | 1 - 2 | 718 | 112 | ||
| AK396604.1 | - | 1 - 2 | 703 | 112 | ||
More Information
Set Membership: |
|

SIAH2
Genomic Map