Assay Details
| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094542.1 | XP_020950201.1 | 1594 | 137 | ||
| XM_021094552.1 | XP_020950211.1 | 1498 | 137 | |||
| GenBank mRNA | AK232627.1 | - | 6 - 7 | 1465 | 137 | |
More Information
Set Membership: |
|

MINDY2
Genomic Map