Assay Details
| Gene Symbol: GJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135162.4 | XP_003135210.1 | 152 | 70 | ||
| GenBank mRNA | AK232707.1 | - | 1 - 2 | 45 | 70 | |
| AK396626.1 | - | 1 - 2 | 65 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135162.4 transcripts.
Set Membership: |
|

GJB1
Genomic Map