Assay Details
| Gene Symbol: UPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929254.6 | XP_001929289.1 | 924 | 59 | ||
| XM_021074095.1 | XP_020929754.1 | 952 | 59 | |||
| GenBank mRNA | AK232712.1 | - | 6 - 7 | 865 | 59 | |
| AK233177.1 | - | 7 - 8 | 997 | 59 | ||
| AK394356.1 | - | 6 - 7 | 895 | 59 | ||
More Information
Set Membership: |
|

UPB1
Genomic Map