Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927695.5 | XP_001927730.1 | 1240 | 68 | ||
| GenBank mRNA | AK232743.1 | - | 4 - 5 | 1234 | 68 | |
More Information
Set Membership: |
|

SLC10A1
Genomic Map