Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021092625.1 | XP_020948284.1 | 290 | 95 | ||
GenBank mRNA | AK232753.1 | - | 2 - 3 | 286 | 95 | |
AK235780.1 | - | 2 - 3 | 270 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
