Assay Details
| Gene Symbol: CPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483537.3 | XP_003483585.1 | 1252 | 61 | ||
| XM_021073052.1 | XP_020928711.1 | 920 | 61 | |||
| GenBank mRNA | AK232759.1 | - | 6 - 7 | 1169 | 61 | |
More Information
Set Membership: |
|

CPN1
Genomic Map