Assay Details
| Gene Symbol: HOMER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927296.5 | XP_001927331.2 | 2190 | 80 | ||
| XM_005666110.3 | XP_005666167.1 | 2383 | 80 | |||
| XM_021098884.1 | XP_020954543.1 | 2221 | 80 | |||
| XM_021098885.1 | XP_020954544.1 | 1872 | 80 | |||
| GenBank mRNA | AK232803.1 | - | 1 | 216 | 80 | |
More Information
Set Membership: |
|

HOMER2
Genomic Map