Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013980506.2 | XP_013835960.2 | 3425 | 102 | ||
| XM_021090342.1 | XP_020946001.1 | 3693 | 102 | |||
| XM_021090343.1 | XP_020946002.1 | 3579 | 102 | |||
| XM_021090344.1 | XP_020946003.1 | 3365 | 102 | |||
| XM_021090345.1 | XP_020946004.1 | 3367 | 102 | |||
| GenBank mRNA | AK232812.1 | - | 1 | 2519 | 102 | |
More Information
Set Membership: |
|

DDAH1
Genomic Map