Assay Details
Gene Symbol: SACS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021065462.1 | XP_020921121.1 | 1392 | 88 | ||
XM_021065463.1 | XP_020921122.1 | 691 | 88 | |||
XM_021065464.1 | XP_020921123.1 | 1392 | 88 | |||
XM_021065465.1 | XP_020921124.1 | 1241 | 88 | |||
XM_021065466.1 | XP_020921125.1 | 1272 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
