Assay Details
| Gene Symbol: SPHK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656915.3 | XP_005656972.1 | 2066 | 53 | ||
| XM_005656916.3 | XP_005656973.1 | 1510 | 53 | |||
| XM_005656918.3 | XP_005656975.1 | 1288 | 53 | |||
| XM_021066589.1 | XP_020922248.1 | 1011 | 53 | |||
| XM_021066590.1 | XP_020922249.1 | 2962 | 53 | |||
| XM_021066591.1 | XP_020922250.1 | 2761 | 53 | |||
| XM_021066592.1 | XP_020922251.1 | 665 | 53 | |||
| XM_021066593.1 | XP_020922252.1 | 595 | 53 | |||
More Information
Set Membership: |
|

SPHK1
Genomic Map