Assay Details
| Gene Symbol: ZNF567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998427.2 | XP_013853881.1 | 1258 | 80 | ||
| XM_021097127.1 | XP_020952786.1 | 882 | 80 | |||
| XM_021097128.1 | XP_020952787.1 | 216 | 80 | |||
| XM_021097129.1 | XP_020952788.1 | 298 | 80 | |||
| XM_021097130.1 | XP_020952789.1 | 292 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013998427.2, XM_021097127.1, XM_021097128.1, XM_021097129.1, XM_021097130.1 transcripts.
Set Membership: |
|

ZNF567
Genomic Map