Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998447.2 | XP_013853901.1 | 502 | 77 | ||
| XM_013998448.2 | XP_013853902.1 | 421 | 77 | |||
| XM_013998449.2 | XP_013853903.1 | 460 | 77 | |||
| XM_013998450.2 | XP_013853904.1 | 685 | 77 | |||
More Information
Set Membership: |
|

ZNF570
Genomic Map