Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998447.2 | XP_013853901.1 | 278 | 85 | ||
| XM_013998448.2 | XP_013853902.1 | 197 | 85 | |||
| XM_013998449.2 | XP_013853903.1 | 236 | 85 | |||
| XM_013998450.2 | XP_013853904.1 | 461 | 85 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013998447.2, XM_013998448.2, XM_013998449.2, XM_013998450.2 transcripts.
Set Membership: |
|

ZNF570
Genomic Map