Assay Details
| Gene Symbol: ZNF570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998447.2 | XP_013853901.1 | 213 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013998447.2 transcripts.
Set Membership: |
|

ZNF570
Genomic Map