Assay Details
| Gene Symbol: LOC102164650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660390.3 | XP_005660447.1 | 772 | 70 | ||
| XM_013993809.2 | XP_013849263.1 | 1082 | 70 | |||
| XM_021068168.1 | XP_020923827.1 | 484 | 70 | |||
| XM_021068169.1 | XP_020923828.1 | 1822 | 70 | |||
More Information
Set Membership: |
|

LOC102164650
Genomic Map