Assay Details
| Gene Symbol: TTBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003353383.5 | XP_003353431.1 | 3738 | 64 | ||
| XM_021097122.1 | XP_020952781.1 | 3401 | 64 | |||
| XM_021097126.1 | XP_020952785.1 | 3860 | 64 | |||
| XM_021097136.1 | XP_020952795.1 | 3637 | 64 | |||
| XM_021097142.1 | XP_020952801.1 | 3238 | 64 | |||
More Information
Set Membership: |
|

TTBK2
Genomic Map